3-Butoxybenzoic acid
Supplier: Matrix Scientific
028189-500MG
101825-654EA
124.62
USD
101825-654
3-Butoxybenzoic acid
3-Butoxybenzoic acid
Hazard: Irritant
Formula:
C₁₁H₁₄O₃ |
MDL Number:
MFCD01143605 CAS Number: 93351-38-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...