3-Ethoxybenzoic acid
Supplier: Matrix Scientific
028186-500MG
101825-648EA
15.38
USD
101825-648
3-Ethoxybenzoic acid
3-Ethoxybenzoic acid
Hazard: Irritant
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol |
MDL Number:
MFCD00016517 CAS Number: 621-51-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...