6-Bromo-2-pyridin-3-ylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

028183-500MG
101825-642EA 146.15 USD
101825-642
6-Bromo-2-pyridin-3-ylquinoline-4-carboxylic acid
6-Bromo-2-pyridin-3-ylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₉BrN₂O₂
MDL Number: MFCD00206126
CAS Number: 5109-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR