6-Ethyl-2-(5-methyl-2-furyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

028177-500MG
101825-630EA 146.15 USD
101825-630
6-Ethyl-2-(5-methyl-2-furyl)quinoline-4-carboxylic acid
6-Ethyl-2-(5-methyl-2-furyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₁₅NO₃
MDL Number: MFCD03422001
CAS Number: 861438-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR