6-Chloro-2-pyridin-2-ylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

028171-500MG
101825-618EA 86.15 USD
101825-618
6-Chloro-2-pyridin-2-ylquinoline-4-carboxylic acid
6-Chloro-2-pyridin-2-ylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₉ClN₂O₂
MDL Number: MFCD03421960
CAS Number: 667412-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR