7,8-Dimethyl-2-pyridin-4-ylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

028162-500MG
101825-600EA 146.15 USD
101825-600
7,8-Dimethyl-2-pyridin-4-ylquinoline-4-carboxylic acid
7,8-Dimethyl-2-pyridin-4-ylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₁₄N₂O₂
MDL Number: MFCD03421952
CAS Number: 588696-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR