8-Chloro-2-(5-chlorothien-2-yl)quinoline-4-carboxylic acid
Supplier: Matrix Scientific
028147-500MG
101825-570EA
146.15
USD
101825-570
8-Chloro-2-(5-chlorothien-2-yl)quinoline-4-carboxylic acid
8-Chloro-2-(5-chlorothien-2-yl)quinoline-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₄H₇Cl₂NO₂S |
MDL Number:
MFCD03422173 CAS Number: 774587-16-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...