8-Chloro-2-(5-methylthien-2-yl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

028141-500MG
101825-558EA 146.15 USD
101825-558
8-Chloro-2-(5-methylthien-2-yl)quinoline-4-carboxylic acid
8-Chloro-2-(5-methylthien-2-yl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₀ClNO₂S
MDL Number: MFCD03422167
CAS Number: 725221-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR