8-Chloro-2-(2,5-dichlorophenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

028133-500MG
101825-542EA 146.15 USD
101825-542
8-Chloro-2-(2,5-dichlorophenyl)quinoline-4-carboxylic acid
8-Chloro-2-(2,5-dichlorophenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₈Cl₃NO₂
MDL Number: MFCD03422159
CAS Number: 863180-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR