2-(4-tert-Butylphenyl)-8-chloroquinoline-4-carboxylic acid
Supplier: Matrix Scientific
028113-500MG
101825-502EA
146.15
USD
101825-502
2-(4-tert-Butylphenyl)-8-chloroquinoline-4-carboxylic acid
2-(4-tert-Butylphenyl)-8-chloroquinoline-4-carboxylic acid
Hazard: Irritant
Formula:
C₂₀H₁₈ClNO₂ |
MDL Number:
MFCD03422140 CAS Number: 862710-11-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...