8-Chloro-2-(4-isopropylphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

028111-500MG
101825-498EA 146.15 USD
101825-498
8-Chloro-2-(4-isopropylphenyl)quinoline-4-carboxylic acid
8-Chloro-2-(4-isopropylphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₉H₁₆ClNO₂
MDL Number: MFCD03422138
CAS Number: 862661-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR