8-Chloro-2-(3-isobutoxyphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

028105-500MG
101825-486EA 146.15 USD
101825-486
8-Chloro-2-(3-isobutoxyphenyl)quinoline-4-carboxylic acid
8-Chloro-2-(3-isobutoxyphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₂₀H₁₈ClNO₃
MDL Number: MFCD03422132
CAS Number: 863185-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR