8-Chloro-2-(4-propoxyphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

028091-500MG
101825-458EA 146.15 USD
101825-458
8-Chloro-2-(4-propoxyphenyl)quinoline-4-carboxylic acid
8-Chloro-2-(4-propoxyphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₉H₁₆ClNO₃
MDL Number: MFCD03422118
CAS Number: 862713-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR