8-Chloro-2-(2-ethoxyphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

028089-500MG
101825-454EA 146.15 USD
101825-454
8-Chloro-2-(2-ethoxyphenyl)quinoline-4-carboxylic acid
8-Chloro-2-(2-ethoxyphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₈H₁₄ClNO₃
MDL Number: MFCD03422116
CAS Number: 862663-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR