8-Chloro-2-(4-ethoxyphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

028085-500MG
101825-446EA 146.15 USD
101825-446
8-Chloro-2-(4-ethoxyphenyl)quinoline-4-carboxylic acid
8-Chloro-2-(4-ethoxyphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₈H₁₄ClNO₃
MDL Number: MFCD03422112
CAS Number: 862663-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR