8-Chloro-2-(3-methoxyphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

028081-500MG
101825-438EA 146.15 USD
101825-438
8-Chloro-2-(3-methoxyphenyl)quinoline-4-carboxylic acid
8-Chloro-2-(3-methoxyphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₁₂ClNO₃
MDL Number: MFCD03422108
CAS Number: 774575-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR