2-(3-Bromophenyl)-8-chloroquinoline-4-carboxylic acid

Supplier: Matrix Scientific

028077-500MG
101825-430EA 146.15 USD
101825-430
2-(3-Bromophenyl)-8-chloroquinoline-4-carboxylic acid
2-(3-Bromophenyl)-8-chloroquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₉BrClNO₂
MDL Number: MFCD03422105
CAS Number: 725217-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR