8-Chloro-2-(3-methylphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

028065-500MG
101825-406EA 146.15 USD
101825-406
8-Chloro-2-(3-methylphenyl)quinoline-4-carboxylic acid
8-Chloro-2-(3-methylphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₁₂ClNO₂
MDL Number: MFCD03422095
CAS Number: 588677-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR