6-Chloro-2-(4-chlorophenyl)-3-methylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

028059-500MG
101825-394EA 146.15 USD
101825-394
6-Chloro-2-(4-chlorophenyl)-3-methylquinoline-4-carboxylic acid
6-Chloro-2-(4-chlorophenyl)-3-methylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₁₁Cl₂NO₂
MDL Number: MFCD03420192
CAS Number: 906143-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR