2-(3-Butoxyphenyl)-6-chloroquinoline-4-carboxylicacid
Supplier: Matrix Scientific
028033-500MG
101825-342EA
146.15
USD
101825-342
2-(3-Butoxyphenyl)-6-chloroquinoline-4-carboxylicacid
2-(3-Butoxyphenyl)-6-chloroquinoline-4-carboxylicacid
Hazard: Irritant
Formula:
C₂₀H₁₈ClNO₃ |
MDL Number:
MFCD03420125 CAS Number: 932841-45-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...