2-(4-Butoxyphenyl)-6-chloroquinoline-4-carboxylic acid

Supplier: Matrix Scientific

028029-500MG
101825-334EA 146.15 USD
101825-334
2-(4-Butoxyphenyl)-6-chloroquinoline-4-carboxylic acid
2-(4-Butoxyphenyl)-6-chloroquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₂₀H₁₈ClNO₃
MDL Number: MFCD03420121
CAS Number: 932841-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR