6-Chloro-2-(2,5-dimethoxyphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

028023-500MG
101825-322EA 215.38 USD
101825-322
6-Chloro-2-(2,5-dimethoxyphenyl)quinoline-4-carboxylic acid
6-Chloro-2-(2,5-dimethoxyphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₈H₁₄ClNO₄
MDL Number: MFCD03420115
CAS Number: 897560-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR