6-Chloro-2-(2-methoxyphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

028019-500MG
101825-314EA 146.15 USD
101825-314
6-Chloro-2-(2-methoxyphenyl)quinoline-4-carboxylic acid
6-Chloro-2-(2-methoxyphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₁₂ClNO₃
MDL Number: MFCD03420112
CAS Number: 895964-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR