2-(4-Butylphenyl)-6-chloroquinoline-4-carboxylic acid
Supplier: Matrix Scientific
028011-500MG
101825-298EA
146.15
USD
101825-298
2-(4-Butylphenyl)-6-chloroquinoline-4-carboxylic acid
2-(4-Butylphenyl)-6-chloroquinoline-4-carboxylic acid
Hazard: Irritant
Formula:
C₂₀H₁₈ClNO₂ |
MDL Number:
MFCD03420106 CAS Number: 932841-37-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...