6-Chloro-2-(5-methyl-2-furyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027995-500MG
101825-266EA 146.15 USD
101825-266
6-Chloro-2-(5-methyl-2-furyl)quinoline-4-carboxylic acid
6-Chloro-2-(5-methyl-2-furyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₀ClNO₃
MDL Number: MFCD03420093
CAS Number: 438216-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR