2-(3-Isopropoxyphenyl)-6,8-dimethylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

027989-500MG
101825-254EA 146.15 USD
101825-254
2-(3-Isopropoxyphenyl)-6,8-dimethylquinoline-4-carboxylic acid
2-(3-Isopropoxyphenyl)-6,8-dimethylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₂₁H₂₁NO₃
MDL Number: MFCD03075315
CAS Number: 438230-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR