2-(2-Ethoxyphenyl)-6,8-dimethylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

027983-500MG
101825-242EA 146.15 USD
101825-242
2-(2-Ethoxyphenyl)-6,8-dimethylquinoline-4-carboxylic acid
2-(2-Ethoxyphenyl)-6,8-dimethylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₂₀H₁₉NO₃
MDL Number: MFCD03420074
CAS Number: 897554-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR