2-(3,4-Dimethoxyphenyl)-6,8-dimethylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

027979-500MG
101825-234EA 146.15 USD
101825-234
2-(3,4-Dimethoxyphenyl)-6,8-dimethylquinoline-4-carboxylic acid
2-(3,4-Dimethoxyphenyl)-6,8-dimethylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₂₀H₁₉NO₄
MDL Number: MFCD03075172
CAS Number: 492997-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR