2-(3-Methoxyphenyl)-6,8-dimethylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

027977-500MG
101825-230EA 146.15 USD
101825-230
2-(3-Methoxyphenyl)-6,8-dimethylquinoline-4-carboxylic acid
2-(3-Methoxyphenyl)-6,8-dimethylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₉H₁₇NO₃
MDL Number: MFCD03075402
CAS Number: 492997-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR