2-(2,4-Dimethylphenyl)-6,8-dimethylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

027962-500MG
101825-200EA 146.15 USD
101825-200
2-(2,4-Dimethylphenyl)-6,8-dimethylquinoline-4-carboxylic acid
2-(2,4-Dimethylphenyl)-6,8-dimethylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₂₀H₁₉NO₂
MDL Number: MFCD03075300
CAS Number: 445260-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR