6,8-Dimethyl-2-(4-methylphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027961-500MG
101825-198EA 146.15 USD
101825-198
6,8-Dimethyl-2-(4-methylphenyl)quinoline-4-carboxylic acid
6,8-Dimethyl-2-(4-methylphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₉H₁₇NO₂
MDL Number: MFCD03420062
CAS Number: 436089-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR