6,8-Dimethyl-2-(3-methylphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027960-500MG
101825-196EA 146.15 USD
101825-196
6,8-Dimethyl-2-(3-methylphenyl)quinoline-4-carboxylic acid
6,8-Dimethyl-2-(3-methylphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₉H₁₇NO₂
MDL Number: MFCD03075086
CAS Number: 438228-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR