6,8-Dimethyl-2-thien-2-ylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

027954-500MG
101825-184EA 200 USD
101825-184
6,8-Dimethyl-2-thien-2-ylquinoline-4-carboxylic acid
6,8-Dimethyl-2-thien-2-ylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₃NO₂S
MDL Number: MFCD03420059
CAS Number: 436092-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR