2-(4-Isobutoxyphenyl)-8-methylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

027945-500MG
101825-166EA 146.15 USD
101825-166
2-(4-Isobutoxyphenyl)-8-methylquinoline-4-carboxylic acid
2-(4-Isobutoxyphenyl)-8-methylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₂₁H₂₁NO₃
MDL Number: MFCD03420043
CAS Number: 438234-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR