2-(4-Isopropoxyphenyl)-8-methylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

027943-500MG
101825-162EA 146.15 USD
101825-162
2-(4-Isopropoxyphenyl)-8-methylquinoline-4-carboxylic acid
2-(4-Isopropoxyphenyl)-8-methylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₂₀H₁₉NO₃
MDL Number: MFCD03074952
CAS Number: 438226-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR