2-(3-Methoxyphenyl)-8-methyl-4-quinolinecarboxylic acid

Supplier: Matrix Scientific

027935-500MG
101825-146EA 109.25 USD
101825-146
2-(3-Methoxyphenyl)-8-methyl-4-quinolinecarboxylic acid
2-(3-Methoxyphenyl)-8-methyl-4-quinolinecarboxylic acid

Hazard: Irritant

Formula: C₁₈H₁₅NO₃
MDL Number: MFCD03075217
CAS Number: 107027-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR