2-(4-sec-Butylphenyl)-8-methylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

027929-500MG
101825-134EA 146.15 USD
101825-134
2-(4-sec-Butylphenyl)-8-methylquinoline-4-carboxylic acid
2-(4-sec-Butylphenyl)-8-methylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₂₁H₂₁NO₂
MDL Number: MFCD03420031
CAS Number: 932796-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR