2-(2,4-Dimethylphenyl)-8-methylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

027922-500MG
101825-120EA 200 USD
101825-120
2-(2,4-Dimethylphenyl)-8-methylquinoline-4-carboxylic acid
2-(2,4-Dimethylphenyl)-8-methylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₉H₁₇NO₂
MDL Number: MFCD03420026
CAS Number: 522596-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR