8-Methyl-2-(2-methylphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027919-500MG
101825-114EA 146.15 USD
101825-114
8-Methyl-2-(2-methylphenyl)quinoline-4-carboxylic acid
8-Methyl-2-(2-methylphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₈H₁₅NO₂
MDL Number: MFCD03420024
CAS Number: 587850-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR