8-Methyl-2-phenylquinoline-4-carboxylic acid
Supplier: Matrix Scientific
027918-500MG
101825-112EA
116.92
USD
101825-112
8-Methyl-2-phenylquinoline-4-carboxylic acid
8-Methyl-2-phenylquinoline-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₇H₁₃NO₂ |
MDL Number:
MFCD00487551 CAS Number: 107027-34-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...