8-Methyl-2-(5-methylthien-2-yl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027915-500MG
101825-106EA 207.69 USD
101825-106
8-Methyl-2-(5-methylthien-2-yl)quinoline-4-carboxylic acid
8-Methyl-2-(5-methylthien-2-yl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₃NO₂S
MDL Number: MFCD03420022
CAS Number: 667412-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR