2-(3-Butoxyphenyl)-6-methylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

027910-500MG
101825-096EA 146.15 USD
101825-096
2-(3-Butoxyphenyl)-6-methylquinoline-4-carboxylic acid
2-(3-Butoxyphenyl)-6-methylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₂₁H₂₁NO₃
MDL Number: MFCD03420012
CAS Number: 932928-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR