6-Methyl-2-(4-propoxyphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027906-500MG
101825-088EA 146.15 USD
101825-088
6-Methyl-2-(4-propoxyphenyl)quinoline-4-carboxylic acid
6-Methyl-2-(4-propoxyphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₂₀H₁₉NO₃
MDL Number: MFCD03074284
CAS Number: 438217-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR