2-(4-Ethoxyphenyl)-6-methylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

027903-500MG
101825-082EA 146.15 USD
101825-082
2-(4-Ethoxyphenyl)-6-methylquinoline-4-carboxylic acid
2-(4-Ethoxyphenyl)-6-methylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₉H₁₇NO₃
MDL Number: MFCD03074548
CAS Number: 438221-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR