2-(4-Bromophenyl)-6-methylquinoline-4-carboxylic acid
Supplier: Matrix Scientific
027898-500MG
101825-072EA
146.15
USD
101825-072
2-(4-Bromophenyl)-6-methylquinoline-4-carboxylic acid
2-(4-Bromophenyl)-6-methylquinoline-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₇H₁₂BrNO₂ |
MDL Number:
MFCD01825278 CAS Number: 351332-55-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...