2-(3-Bromophenyl)-6-methylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

027897-500MG
101825-070EA 146.15 USD
101825-070
2-(3-Bromophenyl)-6-methylquinoline-4-carboxylic acid
2-(3-Bromophenyl)-6-methylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₁₂BrNO₂
MDL Number: MFCD03420008
CAS Number: 725687-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR