2-(4-tert-Butylphenyl)-6-methylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

027893-500MG
101825-062EA 146.15 USD
101825-062
2-(4-tert-Butylphenyl)-6-methylquinoline-4-carboxylic acid
2-(4-tert-Butylphenyl)-6-methylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₂₁H₂₁NO₂
MDL Number: MFCD03074363
CAS Number: 438219-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR