6-Methyl-2-(4-propylphenyl)quinoline-4-carboxylic acid

Supplier: Matrix Scientific

027889-500MG
101825-054EA 146.15 USD
101825-054
6-Methyl-2-(4-propylphenyl)quinoline-4-carboxylic acid
6-Methyl-2-(4-propylphenyl)quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₂₀H₁₉NO₂
MDL Number: MFCD03074361
CAS Number: 438219-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR