2-(2,4-Dimethylphenyl)-6-methylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

027885-500MG
101825-046EA 146.15 USD
101825-046
2-(2,4-Dimethylphenyl)-6-methylquinoline-4-carboxylic acid
2-(2,4-Dimethylphenyl)-6-methylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₉H₁₇NO₂
MDL Number: MFCD03145150
CAS Number: 433243-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR