6-Methyl-2-thien-2-ylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

027878-500MG
101825-032EA 146.15 USD
101825-032
6-Methyl-2-thien-2-ylquinoline-4-carboxylic acid
6-Methyl-2-thien-2-ylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₁NO₂S
MDL Number: MFCD03120601
CAS Number: 31792-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR